3,3,4,4,4-Pentafluoro-1-butyne
Lieferant: Apollo Scientific
|
Gefahr
|
PC450215-5G
APOSPC450215-5GEA
1130
EUR
APOSPC450215-5G
3,3,4,4,4-Pentafluoro-1-butyne
3,3,4,4,4-Pentafluoro-1-butyne
Formel:
C₄HF₅ Molecular Weight: 144,043 g/mol Siedepunkt: –12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18641905 CAS-Nummer: 7096-51-7 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...