4-(Trifluoromethyl)benzaldehyde oxime

Lieferant: Apollo Scientific
Achtung

PC450222-25G
APOSPC450222-25GEA 503 EUR
APOSPC450222-25G
4-(Trifluoromethyl)benzaldehyde oxime
4-(Trifluoromethyl)benzaldehyde oxime
Formel: C₈H₆F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02060973
CAS-Nummer: 66046-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR