4-Iodo-1,1,1-trifluoro-2-(trifluoromethyl)butane 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC450227-1G
APOSPC450227-1GEA
65.4
EUR
APOSPC450227-1G
4-Iodo-1,1,1-trifluoro-2-(trifluoromethyl)butane 98%
4-Iodo-1,1,1-trifluoro-2-(trifluoromethyl)butane
Formel:
C₅H₅F₆I Molecular Weight: 305,988 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16619648 CAS-Nummer: 1346176-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...