5-Chloro-1,1-difluoro-3-oxapentane

Lieferant: Apollo Scientific
Achtung

PC450228-1G PC450228-5G
APOSPC450228-1GEA 405 EUR
APOSPC450228-1G APOSPC450228-5G
5-Chloro-1,1-difluoro-3-oxapentane
5-Chloro-1,1-difluoro-3-oxapentane
Formel: C₄H₇ClF₂O
Molecular Weight: 144.548 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19442046
CAS-Nummer: 1309602-55-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR