Pyridinium trifluoromethanesulphonate
Lieferant: Apollo Scientific
|
Achtung
|
PC450250-100G
PC450250-25G
PC450250-5G
APOSPC450250-100GEA
259
EUR
APOSPC450250-100G
APOSPC450250-25G
APOSPC450250-5G
Pyridinium trifluoromethanesulphonate
Pyridinium trifluoromethanesulphonate
Formel:
C₆H₆F₃NO₃S Schmelzpunkt: 221…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274656 CAS-Nummer: 52193-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...