5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific

PC450256-1G
APOSPC450256-1GEA 575 EUR
APOSPC450256-1G
5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid
5-Nitro-4-(2,2,2-trifluoroacetamido)-2-(trifluoromethyl)benzoic acid
Formel: C₁₀H₄F₆N₂O₅
Molecular Weight: 346,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442153
CAS-Nummer: 1309602-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR