O-(2,2,2-Trifluoroethyl)hydroxylamine hydrochloride 93%
Lieferant: Apollo Scientific
|
Achtung
|
PC450259-250MG
APOSPC450259-250MGEA
139
EUR
APOSPC450259-250MG
O-(2,2,2-Trifluoroethyl)hydroxylamine hydrochloride 93%
O-(2,2,2-Trifluoroethyl)hydroxylamine hydrochloride
Formel:
C₂H₄F₃NO·ClH Molecular Weight: 151,515 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13196692 CAS-Nummer: 68401-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...