Methyl bromochlorofluoroacetate 98%

Lieferant: Apollo Scientific
Achtung

PC450260-1G
APOSPC450260-1GEA 198 EUR
APOSPC450260-1G
Methyl bromochlorofluoroacetate 98%
Methyl bromochlorofluoroacetate
Formel: C₃H₃BrClFO₂
Molecular Weight: 205.41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054291
CAS-Nummer: 101385-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR