Difluoromalonic acid

Lieferant: Apollo Scientific
Gefahr

PC450266-1G
APOSPC450266-1GEA 357 EUR
APOSPC450266-1G
Difluoromalonic acid
Difluoromalonic acid
Formel: C₃H₂F₂O₄
Schmelzpunkt: 117…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11110404
CAS-Nummer: 1514-85-8
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR