(2-Trifluoromethylphenyl)methanesulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Trifluoromethyl)phenyl)methanesulphonamide

PC450275-100G PC450275-25G PC450275-5G
APOSPC450275-100GEA 1080 EUR
APOSPC450275-100G APOSPC450275-25G APOSPC450275-5G
(2-Trifluoromethylphenyl)methanesulphonamide
(2-Trifluoromethylphenyl)methanesulphonamide
Formel: C₈H₈F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09763601
CAS-Nummer: 112941-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR