Benzyl 1,1,2,2-tetrafluoroethyl ether

Lieferant: Apollo Scientific

PC450279-100G PC450279-25G PC450279-500G
APOSPC450279-100GEA 357 EUR
APOSPC450279-100G APOSPC450279-25G APOSPC450279-500G
Benzyl 1,1,2,2-tetrafluoroethyl ether
Benzyl 1,1,2,2-tetrafluoroethyl ether
Formel: C₉H₈F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD16620487
CAS-Nummer: 115611-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR