Bis(2,2,2-trifluoroethyl) Carbonate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Carbonic acid bis(2,2,2-trifluoroethyl) ester
PC450295-25G
APOSPC450295-25GEA
137
EUR
APOSPC450295-25G
Bis(2,2,2-trifluoroethyl) Carbonate
Bis(2,2,2-trifluoroethyl) Carbonate
Formel:
C₅H₄F₆O₃ Molecular Weight: 226.07 g/mol Siedepunkt: 118 °C Dichte: 1.51 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD11857845 CAS-Nummer: 1513-87-7 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...