Hexafluoroisopropyl 2,2,2-trifluoroethyl carbonate
Lieferant: Apollo Scientific
PC450296-25G
APOSPC450296-25GEA
1540
EUR
APOSPC450296-25G
Hexafluoroisopropyl 2,2,2-trifluoroethyl carbonate
Hexafluoroisopropyl 2,2,2-trifluoroethyl carbonate
Formel:
C₆H₃F₉O₃ Molecular Weight: 294,072 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334290 CAS-Nummer: 1980045-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...