2,2,3,3,3-Pentafluoropropyl 2,2,2-trifluoroethyl carbonate
Lieferant: Apollo Scientific
PC450297-25G
APOSPC450297-25GEA
1540
EUR
APOSPC450297-25G
2,2,3,3,3-Pentafluoropropyl 2,2,2-trifluoroethyl carbonate
2,2,3,3,3-Pentafluoropropyl 2,2,2-trifluoroethyl carbonate
Formel:
C₆H₄F₈O₃ Molecular Weight: 276,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334291 CAS-Nummer: 156783-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...