2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450298-25G
APOSPC450298-25GEA 1540 EUR
APOSPC450298-25G
2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate
2,2-Difluoroethyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₆H₅F₇O₃
Molecular Weight: 258,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334292
CAS-Nummer: 1980076-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR