2,2,3,3-Tetrafluoropropyl 2,2,2-trifluoroethyl carbonate
Lieferant: Apollo Scientific
PC450299-25G
APOSPC450299-25GEA
1540
EUR
APOSPC450299-25G
2,2,3,3-Tetrafluoropropyl 2,2,2-trifluoroethyl carbonate
2,2,3,3-Tetrafluoropropyl 2,2,2-trifluoroethyl carbonate
Formel:
C₆H₅F₇O₃ Molecular Weight: 258,091 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334293 CAS-Nummer: 1791423-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...