3-(2,2,2-Trifluoroethoxy)-5-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC45030-1G PC45030-250MG
APOSPC45030-1GEA 0 EUR
APOSPC45030-1G APOSPC45030-250MG
3-(2,2,2-Trifluoroethoxy)-5-(trifluoromethyl)benzoic acid 95%
3-(2,2,2-Trifluoroethoxy)-5-(trifluoromethyl)benzoic acid
Formel: C₁₀H₆F₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD32177069
CAS-Nummer: 2375008-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR