2,2-Difluoroethyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450300-25G
APOSPC450300-25GEA 1540 EUR
APOSPC450300-25G
2,2-Difluoroethyl 2,2,3,3-tetrafluoropropyl carbonate
2,2-Difluoroethyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₆H₆F₆O₃
Molecular Weight: 240,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334294
CAS-Nummer: 1980045-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR