3-Fluoropropyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450301-25G
APOSPC450301-25GEA 1540 EUR
APOSPC450301-25G
3-Fluoropropyl 2,2,2-trifluoroethyl carbonate
3-Fluoropropyl 2,2,2-trifluoroethyl carbonate
Formel: C₆H₈F₄O₃
Molecular Weight: 204,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334295
CAS-Nummer: 1980085-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR