Hexafluoroisopropyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450304-25G
APOSPC450304-25GEA 1540 EUR
APOSPC450304-25G
Hexafluoroisopropyl 2,2,3,3,3-pentafluoropropyl carbonate
Hexafluoroisopropyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₇H₃F₁₁O₃
Molecular Weight: 344.079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334298
CAS-Nummer: 1980038-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR