Bis(2,2,3,3,3-pentafluoropropyl) carbonate
Lieferant: Apollo Scientific
PC450305-25G
APOSPC450305-25GEA
1540
EUR
APOSPC450305-25G
Bis(2,2,3,3,3-pentafluoropropyl) carbonate
Bis(2,2,3,3,3-pentafluoropropyl) carbonate
Formel:
C₇H₄F₁₀O₃ Molecular Weight: 326,089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334299 CAS-Nummer: 154496-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...