2,2,3,3,4,4,4-Heptafluorobutyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450306-25G
APOSPC450306-25GEA 1540 EUR
APOSPC450306-25G
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,2-trifluoroethyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,2-trifluoroethyl carbonate
Formel: C₇H₄F₁₀O₃
Molecular Weight: 326,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334300
CAS-Nummer: 1980044-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR