Hexafluoroisopropyl 2,2,3,3-tetrafluoropropyl carbonate
Lieferant: Apollo Scientific
PC450307-25G
APOSPC450307-25GEA
1540
EUR
APOSPC450307-25G
Hexafluoroisopropyl 2,2,3,3-tetrafluoropropyl carbonate
Hexafluoroisopropyl 2,2,3,3-tetrafluoropropyl carbonate
Formel:
C₇H₄F₁₀O₃ Molecular Weight: 326,089 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334301 CAS-Nummer: 1980039-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...