2,2-Difluoroethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Lieferant: Apollo Scientific
PC450308-25G
APOSPC450308-25GEA
1540
EUR
APOSPC450308-25G
2,2-Difluoroethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
2,2-Difluoroethyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel:
C₇H₅F₉O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334302 CAS-Nummer: 1980048-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...