2,2,3,4,4,4-Hexafluorobutyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450309-25G
APOSPC450309-25GEA 1540 EUR
APOSPC450309-25G
2,2,3,4,4,4-Hexafluorobutyl 2,2,2-trifluoroethyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 2,2,2-trifluoroethyl carbonate
Formel: C₇H₅F₉O₃
Molecular Weight: 308,098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334303
CAS-Nummer: 1980085-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR