2,2,3,3,3-Pentafluoropropyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450310-25G
APOSPC450310-25GEA 1540 EUR
APOSPC450310-25G
2,2,3,3,3-Pentafluoropropyl 2,2,3,3-tetrafluoropropyl carbonate
2,2,3,3,3-Pentafluoropropyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₇H₅F₉O₃
Molecular Weight: 308.098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334304
CAS-Nummer: 1227308-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR