2,2-Difluoroethyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450312-25G
APOSPC450312-25GEA 1540 EUR
APOSPC450312-25G
2,2-Difluoroethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
2,2-Difluoroethyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₇H₆F₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28334305
CAS-Nummer: 1980076-06-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR