3-Fluoropropyl hexafluoroisopropyl carbonate
Lieferant: Apollo Scientific
PC450313-25G
APOSPC450313-25GEA
1540
EUR
APOSPC450313-25G
3-Fluoropropyl hexafluoroisopropyl carbonate
3-Fluoropropyl hexafluoroisopropyl carbonate
Formel:
C₇H₇F₇O₃ Molecular Weight: 272,118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334306 CAS-Nummer: 1980076-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...