3-Fluoropropyl 2,2,3,3,3-pentafluoropropyl carbonate
Lieferant: Apollo Scientific
PC450314-25G
APOSPC450314-25GEA
1540
EUR
APOSPC450314-25G
3-Fluoropropyl 2,2,3,3,3-pentafluoropropyl carbonate
3-Fluoropropyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel:
C₇H₈F₆O₃ Molecular Weight: 254,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334307 CAS-Nummer: 1980044-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...