3-Fluoropropyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450315-25G
APOSPC450315-25GEA 1540 EUR
APOSPC450315-25G
3-Fluoropropyl 2,2,3,3-tetrafluoropropyl carbonate
3-Fluoropropyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₇H₉F₅O₃
Molecular Weight: 236,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334308
CAS-Nummer: 1980048-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR