Bis(3-fluoropropyl) carbonate
Lieferant: Apollo Scientific
PC450316-25G
APOSPC450316-25GEA
1540
EUR
APOSPC450316-25G
Bis(3-fluoropropyl) carbonate
Bis(3-fluoropropyl) carbonate
Formel:
C₇H₁₂F₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334309 CAS-Nummer: 1132677-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...