2,2,3,3,4,4,4-Heptafluorobutyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450317-25G
APOSPC450317-25GEA 1540 EUR
APOSPC450317-25G
2,2,3,3,4,4,4-Heptafluorobutyl hexafluoroisopropyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl hexafluoroisopropyl carbonate
Formel: C₈H₃F₁₃O₃
Molecular Weight: 394,087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334310
CAS-Nummer: 1980035-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR