2,2,3,4,4,4-Hexafluorobutyl hexafluoroisopropyl carbonate

Lieferant: Apollo Scientific

PC450318-25G
APOSPC450318-25GEA 1540 EUR
APOSPC450318-25G
2,2,3,4,4,4-Hexafluorobutyl hexafluoroisopropyl carbonate
2,2,3,4,4,4-Hexafluorobutyl hexafluoroisopropyl carbonate
Formel: C₈H₄F₁₂O₃
Molecular Weight: 376,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334311
CAS-Nummer: 1980038-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR