2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450319-25G
APOSPC450319-25GEA 1540 EUR
APOSPC450319-25G
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₈H₄F₁₂O₃
Molecular Weight: 376,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334312
CAS-Nummer: 1980085-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR