2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450320-25G
APOSPC450320-25GEA 1540 EUR
APOSPC450320-25G
2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₈H₅F₁₁O₃
Molecular Weight: 358.106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334313
CAS-Nummer: 1980039-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR