2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450321-25G
APOSPC450321-25GEA 1540 EUR
APOSPC450321-25G
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
2,2,3,3,4,4,4-Heptafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₈H₅F₁₁O₃
Molecular Weight: 358,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334314
CAS-Nummer: 1980048-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR