2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450322-25G
APOSPC450322-25GEA 1540 EUR
APOSPC450322-25G
2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
2,2,3,4,4,4-Hexafluorobutyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₈H₆F₁₀O₃
Molecular Weight: 340,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334315
CAS-Nummer: 1980085-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR