3-Fluoropropyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450324-25G
APOSPC450324-25GEA 1540 EUR
APOSPC450324-25G
3-Fluoropropyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
3-Fluoropropyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formel: C₈H₈F₈O₃
Molecular Weight: 304,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334317
CAS-Nummer: 1980048-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR