3-Fluoropropyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Lieferant: Apollo Scientific

PC450325-25G
APOSPC450325-25GEA 1540 EUR
APOSPC450325-25G
3-Fluoropropyl 2,2,3,4,4,4-hexafluorobutyl carbonate
3-Fluoropropyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formel: C₈H₉F₇O₃
Molecular Weight: 286,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334318
CAS-Nummer: 1980076-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR