2,2-Difluoroethyl 4,4,5,5,5-pentafluoropentyl carbonate
Lieferant: Apollo Scientific
PC450326-25G
APOSPC450326-25GEA
1540
EUR
APOSPC450326-25G
2,2-Difluoroethyl 4,4,5,5,5-pentafluoropentyl carbonate
2,2-Difluoroethyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel:
C₈H₉F₇O₃ Molecular Weight: 286,144 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334319 CAS-Nummer: 1980040-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...