Bis(2,2,3,3,4,4,4-heptafluorobutyl) carbonate
Lieferant: Apollo Scientific
PC450327-25G
APOSPC450327-25GEA
1540
EUR
APOSPC450327-25G
Bis(2,2,3,3,4,4,4-heptafluorobutyl) carbonate
Bis(2,2,3,3,4,4,4-heptafluorobutyl) carbonate
Formel:
C₉H₄F₁₄O₃ Molecular Weight: 426,104 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334320 CAS-Nummer: 1026739-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...