2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450328-25G
APOSPC450328-25GEA
1540
EUR
APOSPC450328-25G
2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate
2,2,2-Trifluoroethyl 1H,1H-perfluorohexyl carbonate
Formel:
C₉H₄F₁₄O₃ Molecular Weight: 426,104 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334321 CAS-Nummer: 1980076-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...