2,2-Difluoroethyl 1H,1H-perfluorohexyl carbonate
Lieferant: Apollo Scientific
PC450329-25G
APOSPC450329-25GEA
1540
EUR
APOSPC450329-25G
2,2-Difluoroethyl 1H,1H-perfluorohexyl carbonate
2,2-Difluoroethyl 1H,1H-perfluorohexyl carbonate
Formel:
C₉H₅F₁₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334322 CAS-Nummer: 1980035-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...