Bis(2,2,3,4,4,4-hexafluorobutyl) carbonate
Lieferant: Apollo Scientific
PC450331-25G
APOSPC450331-25GEA
1540
EUR
APOSPC450331-25G
Bis(2,2,3,4,4,4-hexafluorobutyl) carbonate
Bis(2,2,3,4,4,4-hexafluorobutyl) carbonate
Formel:
C₉H₆F₁₂O₃ Molecular Weight: 390,123 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28334324 CAS-Nummer: 847755-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...