1H,1H,2H,2H-Perfluorohexyl 2,2,2-trifluoroethyl carbonate

Lieferant: Apollo Scientific

PC450332-25G
APOSPC450332-25GEA 1540 EUR
APOSPC450332-25G
1H,1H,2H,2H-Perfluorohexyl 2,2,2-trifluoroethyl carbonate
1H,1H,2H,2H-Perfluorohexyl 2,2,2-trifluoroethyl carbonate
Formel: C₉H₆F₁₂O₃
Molecular Weight: 390,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334325
CAS-Nummer: 1980044-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR