2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate

Lieferant: Apollo Scientific

PC450333-25G
APOSPC450333-25GEA 1540 EUR
APOSPC450333-25G
2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate
2,2-Difluoroethyl 1H,1H,2H,2H-perfluorohexyl carbonate
Formel: C₉H₇F₁₁O₃
Molecular Weight: 372,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334326
CAS-Nummer: 1980048-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR