Hexafluoroisopropyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450334-25G
APOSPC450334-25GEA 1540 EUR
APOSPC450334-25G
Hexafluoroisopropyl 4,4,5,5,5-pentafluoropentyl carbonate
Hexafluoroisopropyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₉H₇F₁₁O₃
Molecular Weight: 372,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334327
CAS-Nummer: 1980086-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR