4,4,5,5,5-Pentafluoropentyl 2,2,3,3,3-pentafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450335-25G
APOSPC450335-25GEA 1540 EUR
APOSPC450335-25G
4,4,5,5,5-Pentafluoropentyl 2,2,3,3,3-pentafluoropropyl carbonate
4,4,5,5,5-Pentafluoropentyl 2,2,3,3,3-pentafluoropropyl carbonate
Formel: C₉H₈F₁₀O₃
Molecular Weight: 354,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334328
CAS-Nummer: 1980085-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR