4,4,5,5,5-Pentafluoropentyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific

PC450336-25G
APOSPC450336-25GEA 1540 EUR
APOSPC450336-25G
4,4,5,5,5-Pentafluoropentyl 2,2,3,3-tetrafluoropropyl carbonate
4,4,5,5,5-Pentafluoropentyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₉H₉F₉O₃
Molecular Weight: 336,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334329
CAS-Nummer: 1980039-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR