3-Fluoropropyl 4,4,5,5,5-pentafluoropentyl carbonate

Lieferant: Apollo Scientific

PC450337-25G
APOSPC450337-25GEA 1540 EUR
APOSPC450337-25G
3-Fluoropropyl 4,4,5,5,5-pentafluoropentyl carbonate
3-Fluoropropyl 4,4,5,5,5-pentafluoropentyl carbonate
Formel: C₉H₁₂F₆O₃
Molecular Weight: 282,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28334330
CAS-Nummer: 1980038-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR